Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LONP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549215

Catalog No. PA549215


Add to cart

Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

ATP-dependent serine protease that mediates the selective degradation of misfolded, unassembled or oxidatively damaged polypeptides as well as certain short-lived regulatory proteins in the mitochondrial matrix. May also have a chaperone function in the assembly of inner membrane protein complexes. Participates in the regulation of mitochondrial gene expression and in the maintece of the integrity of the mitochondrial genome. Binds to mitochondrial promoters and RNA in a single- stranded, site-specific, and strand-specific manner. May regulate mitochondrial DNA replication and/or gene expression using site- specific, single-stranded DNA binding to target the degradation of regulatory proteins binding to adjacent sites in mitochondrial promoters. Endogenous substrates include mitochondrial steroidogenic acute regulatory (StAR) protein.
Specifications

Specifications

LONP1
Polyclonal
PBS with 0.09% sodium azide
P36776
Lon protease homolog, mitochondrial, 3421-, LONHs, Lon protease-like protein, LONP, Mitochondrial ATP-dependent protease Lon, Serine protease 15, LONP1, PRSS15
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9361
Western Blot
Unconjugated
LONP1
Liquid
LONP1
KLH conjugated synthetic peptide between 389-418 amino acids from the Central region of human LONP1.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit