Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuGENE™ 10X Tris-Glycine-SDS Buffers, Lonza™

For use with denaturing protein electrophoresis

$194.90 - $889.00

Specifications

Concentration 10X
Formulation Tris Glycine-SDS Buffer, 10X with 0.25M Tris base, 1.92M Glycine, 1% SDS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BMA50880
View Documents Promotion Details
Lonza IncSupplier Diversity Partner
50880
1 L
Each for $194.90
Add to cart
 
BMA50882
View Documents Promotion Details
Lonza IncSupplier Diversity Partner
50882
4 L
Each for $889.00
Add to cart
 
Description

Description

  • Gold standard
  • Formulated for use with PAGEr™ precast gels
  • Clear or colorless aqueous solution
  • Miscible in water
  • Prepared with high quality reagents and use 18megohm water
  • Filtered using 0.2μ filter
  • Ready-to-use

Specifications

Specifications

10X
Tris Glycine-SDS Buffer, 10X with 0.25M Tris base, 1.92M Glycine, 1% SDS
Safety and Handling

Safety and Handling

Recommended Storage : 18° to 26°C

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit