Learn More
Lonza™ AccuGENE™ 20X SSC Buffer
Supplier:  Lonza™ 51205
Catalog No. BMA51205
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
AccuGENE Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications. Each reagent is prepared with 18megOhm water, filtered using a 0.2 micron filter and placed into sterile bottles.
  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency.
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted.
  • Efficient - Ready-made solutions eliminate experiment preparation time.
  • Flexible - Customized solutions are available to meet individual needs
Specifications
Specifications
SSC Buffer
Sterile
7
1 L
Product Suggestions
Product Suggestions