Learn More
AccuGENE™ TAE Buffers, Lonza™
Formulated to match Lonza's Reliant™ and Latitude™ gels
$151.50 - $590.85
Catalog Number | Mfr. No. | Concentration | Quantity | Formulation | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Quantity | Formulation | Price | Quantity & Availability | |||
BMA50844
|
Lonza Inc
50844 |
10X | 1 L | TAE Buffer, 10X with 0.4M Tris acetate, 0.01M EDTA |
Each for $151.50
|
|
|||
BMA50841
|
Lonza Inc
50841 |
10X | 4 L | TAE Buffer, 10X with 0.4M Tris acetate, 0.01M EDTA |
Each for $590.85
|
|
|||
BMA51216
|
Lonza Inc
51216 |
50X | 1 L | TAE Buffer, 50X with 2M Tris acetate, 0.05M EDTA |
Each for $310.07
|
|
|||
Description
- Ready-to-use
- Prepared with high quality reagents and use 18megohm water
- Filtered using 0.2μ filter
- Available in 10X and 50X strength
- Manufactured according to strict quality control standards to ensure consistency
- Customized solutions are available to meet individual needs
Safety and Handling
Recommended Storage : 18° to 26°C