Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ 293 Cell Serum-Free Media

Completely balanced to support vaccine and viral vector production of high-density human embryonic kidney (HEK) 293 cell culture environments

$134.90

Specifications

Without Additives L-Glutamine and Phenol Red
Packaging Bottle
Quantity 1L
Storage Requirements 2° to 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
BW12-765Q
View Documents
Lonza™
12765Q
Pro293S-CDM, for suspension cells
Each for $134.90
Add to cart
 
BW12-764Q
View Documents
Lonza™
12764Q
Pro293A-CDM, for adherent cells This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Very low levels of recombinant proteins facilitate downstream purification and provide regulatory compliance

  • Chemically-defined media optimized for specific cell types

† Store as detailed on label.

Specifications

Specifications

L-Glutamine and Phenol Red
Bottle
1L
2° to 8°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit