Learn More
Lonza™ BioWhittaker™ Antibiotics: Gentamicin Sulfate
Used as a broad spectrum bactericidal, with activity against gram negative and gram positive organisms including Pseudomonas.   BioWhittaker™ Antibiotics: Gentamicin Sulfate is manufactured in accordance with cGMP regulations and cell culture tested to assure compatibility.
Manufacturer:  Lonza™ 17528Z
 View more versions of this product
Catalog No. BW17-528Z
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Broad spectrum bactericida
  • Stable over a wide range of pH and temperature
  • Autoclavable
  • Colorless to amber or yellow.
  • cGMP compatibility
Specifications
Specifications
50mg/mL
15° to 30°C
10mL
3.5 to 5.5
Crimp top vial
15° to 30°C
Gentamicin Sulfate