Learn More
Lonza™ BioWhittaker™ Antibiotics: Penicillin-Streptomycin Mixtures
Used as a broad spectrum bacteriostatic and bactericidal, with activity against gram negative and gram-positive organisms including yeast and mold. BioWhittaker™ Antibiotics: Penicillin-Streptomycin Mixtures are manufactured in accordance with cGMP regulations.
Supplier:  Lonza™ 17745E
 View more versions of this product
Catalog No. BW17-745E
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
This item is not returnable. View return policy
Description
Description
  • Suggested working concentration of Penicillin-Streptomycin is 100 units of Potassium Penicillin and 100 μg of Streptomycin Sulfate per 1 ml of culture media
  • Manufactured in accordance with cGMP regulations
  • Cell culture tested to assure compatibility.
  • 20mL vials have crimp-top closure
  • 4.5, 100 and 500mL bottles have screw-cap closure
Specifications
Specifications
-10° to -20°C
10K/10K/25μg
100 mL
Penicillin-Streptomycin-Fungizone
3.3 to 7.2