Learn More
Lonza™ BioWhittaker™ Trypsin Reagent
Used to remove cells from the culture vessel surface into suspension.  BioWhittaker™ Trypsin Reagentis porcine parvovirus tested and mycoplasma screened
Manufacturer:  Lonza™ 17160E
 View more versions of this product
Catalog No. BW17-160E
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Clear, colorless-yellow
  • Porcine enzyme preparation, used to remove cells from the culture vessel surface into suspension. It is Generally used at a working concentration of 0.25%
  • Prepared in a balanced salt solution, without calcium or magnesium at a pH of 7.6-7.8
  • Trypsin activity can be halted by adding a serum media mixture or a Trypsin inhibitor
  • Porcine parvovirus tested and mycoplasma screened
  • Product is manufactured with irradiated trypsin
Specifications
Specifications
-10° to -20°C
100 mL
6.44 to 8.04
Sterile
Calcium, Magnesium