Learn More
Lonza™ BioWhittaker™ Trypsin Reagent
Used to remove cells from the culture vessel surface into suspension.  BioWhittaker™ Trypsin Reagentis porcine parvovirus tested and mycoplasma screened
Manufacturer:  Lonza™ 17161F
 View more versions of this product
Catalog No. BW17-161F
Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Clear, colorless-yellow
  • Porcine enzyme preparation, used to remove cells from the culture vessel surface into suspension. It is Generally used at a working concentration of 0.25%
  • Prepared in a balanced salt solution, without calcium or magnesium at a pH of 7.6-7.8
  • Trypsin activity can be halted by adding a serum media mixture or a Trypsin inhibitor
  • Porcine parvovirus tested and mycoplasma screened
  • Product is manufactured with irradiated trypsin
Specifications
Specifications
-10° to -20°C
Sterile
500 mL
7.1 to 7.9