Learn More
Lonza BioWhittaker Trypsin Reagent
Used to remove cells from the culture vessel surface into suspension. BioWhittaker Trypsin Reagentis porcine parvovirus tested and mycoplasma screened
$86.00 - $345.00
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity & Availability | ||||
BW17-160E
|
Lonza
17160E |
Trypsin(10x) 2.5% in HBSS w/o Ca, Mg | 100mL |
Each for $86.00
|
|
BW17-160F
|
Lonza
17160F |
Trypsin(10x) 2.5% in HBSS w/o Ca, Mg | 500mL |
Each for $345.00
|
|
BW17-161F
|
Lonza
17161F |
Trypsin-Versene™ (EDTA) | 500mL |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
Description
- Clear, colorless-yellow
- Porcine enzyme preparation, used to remove cells from the culture vessel surface into suspension. It is Generally used at a working concentration of 0.25%
- Prepared in a balanced salt solution, without calcium or magnesium at a pH of 7.6-7.8
- Trypsin activity can be halted by adding a serum media mixture or a Trypsin inhibitor
- Porcine parvovirus tested and mycoplasma screened
- Product is manufactured with irradiated trypsin
Specifications
Sterile |