Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ CHO (Chinese Hamster Ovary) Serum-Free Culture Media: ProCHO4

For optimized expression of protein in CHO cells

Manufacturer:  Lonza™ 04919Q

 View more versions of this product

Catalog No. BW04919Q


Add to cart
This item is not returnable. View return policy

Description

Description

  • ProCHO-4 is designed for the concurrent transition of adherent CHO cells to serum-free and suspension culture; supports faster doubling times

†Store as detailed on label.

Specifications

Specifications

2° to 8°C
Liquid
None
1L
CHO
Phenol Red, 0.1% Pluronic F-68, L-Glutamine, Hypoxanthine or Thymidine
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit