Learn More
Lonza™ BioWhittaker™ General-Purpose Serum-Free Media
Chemically defined media optimized for specific cell types
Manufacturer:  Lonza™ 344022
 View more versions of this product
Catalog No. BW344022
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PC-1™
  • Provides complete media system, including PC-1 Supplement for cultivation of primary cells and anchorage-dependent cell lines
  • Contains complete HEPES buffering system
UltraCULTURE™
  • Designed for growth of adherent and nonadherent cells
Specifications
Specifications
PC-1™ medium Supplement
2° to 8°C
10mL