Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ General-Purpose Serum-Free Media

Chemically defined media optimized for specific cell types

$140.80 - $402.00

Specifications

Storage Requirements 2° to 8°C
View More Specs

Products 3
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
BW77232
View Documents
Lonza™
77232
PC-1™ CD Serum-free medium 2 x 500mL Pack of 2 for $402.00
Add to cart
 
BW344022
View Documents
Lonza™
344022
PC-1™ medium Supplement 10mL Each for $140.80
Add to cart
 
BW12-725F
View Documents
Lonza™
12725F
UltraCULTURE™ Serum-free medium 500mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

PC-1™

  • Provides complete media system, including PC-1 Supplement for cultivation of primary cells and anchorage-dependent cell lines
  • Contains complete HEPES buffering system
UltraCULTURE™
  • Designed for growth of adherent and nonadherent cells

Specifications

Specifications

2° to 8°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit