Learn More
Lonza™ BioWhittaker™ Hank's Balanced Salt Solutions (1X)
Used in wide variety of tissue culture applications. BioWhittaker™ Hank's Balanced Salt Solutions (1X) is used to maintain a physiological pH for cells maintained in non-CO2 conditions, diluting cells before counting, washing cells before dissociating and transporting cells or tissues.
Manufacturer:  Lonza™ 10508Q
 View more versions of this product
Catalog No. BW10-508Q
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Available in various formulations including calcium-, magnesium- and phenol-red-free or containing versions
  • Manufactured in accordance with cGMP regulations and are cell culture tested to assure compatibility
  • Contain NaHCO3
Specifications
Specifications
Hank's balanced salt solution
Sterile
With Phenol Red
7.1 to 7.4
15° to 30°C
1L
Phenol Red