Learn More
Lonza™ BioWhittaker™ Hybridoma Serum-Free and Protein-Free Culture Media: HL-1 Supplement (100X)
Prescreened in actual hybridoma systems to ensure minimal variation
Manufacturer:  Lonza™ 77227
Catalog No. BW77227
Add to cart
Description
Description
  • HL-1 supports the serum-free growth of hybridomas and differentiated cells of lymphoid origin
1Rehydrates to quantity
Specifications
Specifications
Liquid
10mL
2° to 8°C