Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lonza™ BioWhittaker™ Insect Cell Culture Media: Grace's Insect Medium
Chemically-defined media optimized for specific cell types
Supplier: Lonza™ 04457F
This item is not returnable.
View return policy
Description
Compliance
Manufactured in accordance with cGMP regulations for insect cell lines; formulations recommended by the Tissue Culture Association.
Specifications
2° to 8°C | |
Grace's Insect Medium | |
No Lactalbumin Hydrolysate, No Yeastolate, No Gentamicin |
Liquid | |
None | |
500 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title