Learn More
Lonza™ BioWhittaker Insect-XPRESS Insect Cell Culture Media (1X) with L-Glutamine
Designed to support the growth of insect cell lines derived from Spodoptera frugiperda (Sf9 and Sf21). BioWhittaker™ Insect-XPRESS™ Insect Cell Culture Media (1X) with L-Glutamine supports superior production of recombinant proteins when using the Baculovirus Expression Vector System (BEVS).
Supplier:  Lonza™ 12730Q
 View more versions of this product
Catalog No. BW12-730Q
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Protein-free
  • Cell densities in excess of 8.3×106 cells/mL can be achieved with suspension cultures of Sf9 cells using Insect-XPRESS Medium and excess of oxygen
  • Formulation can also be used for stationary mono¬layer cultures and shake-flask cultures
Compliance
Manufactured in accordance with cGMP regulations for insect cell lines; formulations recommended by the Tissue Culture Association.
Specifications
Specifications
2° to 8°C
Insect Cell Medium, Protein-free
1 L
Liquid
Glutamine