Learn More
Lonza™ BioWhittaker™ Iscove's Modified Dulbecco's Medium with L-Glutamine
Manufacturer:  Lonza™ 12915F
 View more versions of this product
Catalog No. BW12-915F
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Sterile-filtered through two consecutive 0.2μm pharmaceutical-grade filters
2° to 8°C
500mL
IMDM (Screened to Support Hybridoma Growth)
6.95 to 7.15
2° to 8°C
Sterile
Liquid
L-Glutamine and 25mM HEPES Buffer