Learn More
Lonza™ BioWhittaker™ L-Glutamine (200mM)
Manufactured in accordance with cGMP regulations
Manufacturer:  Lonza™ 17605C
 View more versions of this product
Catalog No. BW17-605C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Required for cell culture growth
  • Normal concentration of L-glutamine in media is 292mg/L or 2mM
  • L-glutamine is not stable for long periods of time unless it is frozen
  • Material should be aliquoted to smaller volumes and then frozen
  • Stability of L-glutamine in culture medium is primarily dependent on the temperature at which it is stored
  • Lonza test data for long-term storage indicates that the L-glutamine concentration is reduced by approximately 2 to 3% per month in tissue culture medium stored at 2° to 8°C
  • Basal medium containing L-glutamine supports cell growth for up to 24 months when stored in refrigerated conditions
Specifications
Specifications
L-glutamine
Sterile
Liquid
-10° to -20°C
50mL
5.58 to 6.78