Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ MEM Eagle with Hanks' BSS

Supplier:  Lonza™ 12137F

 View more versions of this product

Catalog No. BW12-137F

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

MEM Eagle is suitable for a diverse spectrum of mammalian cell types.

Specifications

Specifications

15° to 30°C
Sterile
No Glutamine
500 mL
MEM Eagle/HBSS
Sterile-filtered through two consecutive 0.2 μm filters
7.02 to 7.42
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit