Learn More
Lonza™ BioWhittaker™ PowerCHO™ CD Medium
Support the growth of CHO K1, DXB11, DG44 and GS systems
Manufacturer:  Lonza™ 12770Q
 View more versions of this product
Catalog No. BW12770Q
Add to cart
Description
Description
PowerCHO 1, 2, and 3 Chemically Defined, Serum-free CHO Media are the next generation in CHO media, optimized for both cell growth and protein production. For therapeutic bioprocessing applications, these protein-free formulations also facilitate both downstream purification and regulatory compliance.
  • Non-animal origin media
  • Hydrolysate-free, serum-free
  • Contains HEPES and Pluronic™ F-68
  • Free of glutamine, hypoxanthine, thymidine and phenol red
  • Maintain high viability (>90%) at high cell densities
  • Support high-density, large scale production volumes through balanced formulation
Specifications
Specifications
2° to 8°C
1L
PowerCHO 1
L-Glutamine, Phenol Red, Hypoxanthine and Thymidine