Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lonza™ BioWhittaker™ PowerCHO™ CD Medium
Support the growth of CHO K1, DXB11, DG44 and GS systems
Specifications
Content And Storage | 2° to 8°C |
---|---|
Cell Line | CHO |
Without Additives | No Glutamine, No Phenol Red, No Hypoxanthine, No Thymidine |
Quantity | 1 L |
Description
PowerCHO 1, 2, and 3 Chemically Defined, Serum-free CHO Media are the next generation in CHO media, optimized for both cell growth and protein production. For therapeutic bioprocessing applications, these protein-free formulations also facilitate both downstream purification and regulatory compliance.
- Non-animal origin media
- Hydrolysate-free, serum-free
- Contains HEPES and Pluronic™ F-68
- Free of glutamine, hypoxanthine, thymidine and phenol red
- Maintain high viability (>90%) at high cell densities
- Support high-density, large scale production volumes through balanced formulation
Specifications
2° to 8°C | |
CHO | |
No Glutamine, No Phenol Red, No Hypoxanthine, No Thymidine | |
1 L |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title