Learn More
Lonza™ BioWhittaker™ ProVero 1 Kidney Cell Medium
Optimized for the Vero renal expression model.
Manufacturer:  Lonza™ BE02030Q
Catalog No. BWBE02030Q
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
ProVero™ 1 Serum-free Medium is a non-animal origin, protein-free medium designed to support the growth of MDCK and vero cells. ProVero™ 1 Medium includes HEPES and sodium bicarbonate buffer. The absence of proteins and only very low levels of human recombinant insulin facilitate both downstream processing and regulatory compliance. Some vero cell strains require additional supplementation with 5.0μg/L rhEGF for optimal vero cell growth.
  • Recombinant protein production
  • Virus production
Specifications
Specifications
ProVero™ 1 Serum-free medium
2° to 8°C
1L