Learn More
Lonza™ FlashGel™ System Sample Preparation Reagents
For use with the Lonza FlashGel System.  FlashGel™ System Sample Preparation Reagents for the FlashGel™ System provide the fastest separations and enable the user to watch as nucleic acids migrate in the gel.
$254.00 - $256.00
Specifications
Shelf Life 12 Months
Storage Requirements Room Temperature
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BMA57060
View Documents
Lonza™
57060
2 x 500 μL
Each for $254.00
Add to cart
BMA50462
View Documents
Lonza™
50462
5 x 1 mL
Each for $256.00
Add to cart
Description
Description
  • Reagents include Recovery Buffer and Loading Dye
  • Loading Dye: 5X concentration; RNA native sample buffer
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
Specifications
Specifications
12 Months
Room Temperature