Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ Latitude™ Midigel Chamber

Provides optimal running conditions for Latitude Precast Midigels

Manufacturer:  Lonza™ 56990

Catalog No. BMA56990

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: Acrylic buffer chamber and Acrylic safety cover with attached power supply leads.

Description

Description

  • Chamber has a red/orange background for easy sample loading
  • Compact buffer compartment minimizes buffer usage

Note:this unit is designed for use with Latitude Precast Gels; casting accessories are not available

Specifications

Specifications

200 mA
Latitude™ Midigel Chamber
7 cm
9.4 in.
Acrylic
250 VDC
350 to 400 mL
15 w
11.8 cm
9.4 x 4.6 x 2.7 in. (24 x 11.8 x 7 cm)
2.7 in.
Acrylic buffer chamber and Acrylic safety cover with attached power supply leads.
24 cm
55°C
11.8 to 13.5 oz.
1 Year
4.6 in.
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit