Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ PAGEr™ EX Protein Stain Kits

Designed for separation and staining in less than 30 minutes

Specifications

Format 12-well, 9 x 10 cm
For Use With (Application) Designed for separation and staining in less than 30 min.
Kit Contents 2 PAGEr™ EX Gels, 100mL ProSieve™ EX Running Buffer, 25mL ProSieve™ EX Safe Stain, 50μL ProSieve™ Quad Color Marker
Product Type Protein Stain Kit
View More Specs

Includes:
  • 2 PAGEr™ EX Gels
  • 100mL ProSieve™ EX Running Buffer
  • 25mL ProSieve™ EX Safe Stain
  • 50µL ProSieve™ Quad Color Marker

Products 4
Catalog Number Mfr. No. Separation Range Price Quantity & Availability  
Catalog Number Mfr. No. Separation Range Price Quantity & Availability  
BMA00201741
View Documents
Lonza™
00201741
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
BMA00201745
View Documents
Lonza™
00201745
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
BMA00201746
View Documents
Lonza™
00201746
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
BMA00201748
View Documents
Lonza™
00201748
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • 10-minute, non-toxic (safe) stain with only 1 step
  • 20-minute, run/separation

Specifications

Specifications

12-well, 9 x 10 cm
Designed for separation and staining in less than 30 min.
2 PAGEr™ EX Gels, 100mL ProSieve™ EX Running Buffer, 25mL ProSieve™ EX Safe Stain, 50μL ProSieve™ Quad Color Marker
Protein Stain Kit
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit