Learn More
Lonza™ PAGEr™ EX Protein Transfer Blotting Kits
Designed for separation and staining in less than 30 minutes
Manufacturer:  Lonza™ 00201744
 View more versions of this product
Catalog No. BMA00201744
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes:
  • 2 PAGEr™ EX Gels
  • 100mL ProSieve™ EX Running Buffer
  • 25mL ProSieve™ EX Safe Stain
  • 50µL ProSieve™ Quad Color Marker
Description
Description
  • 10-minute non-toxic (safe) stain with only 1 step
  • 20-minute run/separation
Specifications
Specifications
Designed for separation and staining in less than 30 min.
2 PAGEr™ EX Gels, 100mL ProSieve™ EX Running Buffer, 25mL ProSieve™ EX Safe Stain, 50μL ProSieve™ Quad Color Marker
10 cm
5 to 225 kd
1 Kit: 2 PAGEr™ EX Gels, 100 mL ProSieve™ EX Running Buffer, 25 mL ProSieve™ EX Safe Stain, 50 &mu:L ProSieve™ QuadColor™ Marker
12-well, 10 x 10 cm
10 cm
Protein Transfer/Western Blot Kit
12-well
Safety and Handling
Safety and Handling
ShelfLife : 12 Months