Learn More
Lonza™ PAGEr™ EX Protein Transfer Blotting Kits
Designed for separation and staining in less than 30 minutes
Specifications
For Use With (Application) Designed for separation and staining in less than 30 min.
Kit Contents 2 PAGEr™ EX Gels, 100mL ProSieve™ EX Running Buffer, 25mL ProSieve™ EX Safe Stain, 50μL ProSieve™ Quad Color Marker
Product Type Protein Transfer/Western Blot Kit
View More Specs
Includes:
  • 2 PAGEr™ EX Gels
  • 100mL ProSieve™ EX Running Buffer
  • 25mL ProSieve™ EX Safe Stain
  • 50µL ProSieve™ Quad Color Marker
Products 4
Catalog Number Mfr. No. Separation Range Price Quantity & Availability  
Catalog Number Mfr. No. Separation Range Price Quantity & Availability  
BMA00201742
View Documents
Lonza™
00201742
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
BMA00201743
View Documents
Lonza™
00201743
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
BMA00201744
View Documents
Lonza™
00201744
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
BMA00201747
View Documents
Lonza™
00201747
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • 10-minute non-toxic (safe) stain with only 1 step
  • 20-minute run/separation
Specifications
Specifications
Designed for separation and staining in less than 30 min.
2 PAGEr™ EX Gels, 100mL ProSieve™ EX Running Buffer, 25mL ProSieve™ EX Safe Stain, 50μL ProSieve™ Quad Color Marker
Protein Transfer/Western Blot Kit
Safety and Handling
Safety and Handling
ShelfLife : 12 Months