Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ PAGEr™ Gold 4 to 20%, Tris-Glycine, Mini, Protein Gel, 12-well

Separation Range: 5 to 200 kd; Separation Type: Denaturing, Native; For Use With: Hoefer Mighty Small SE 250

Manufacturer:  Lonza™ 59505

Catalog No. BMA59505

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PAGEr Gold Precast Gels are easy-to-use protein minigels that offer sharp resolution and consistent protein transfer. Choose from multiple options for denatured or native protein separation over a wide molecular weight range, in an array of well configurations in both 9 × 10cm (see BMA58517 through BMA58524) and 10 × 10cm sizes to fit most popular chambers.

  • Polyacrylamide minigels cast with 4% stacking gel for vertical protein electrophoresis
  • Provide easy, accurate separation of biomolecules over wide molecular-weight range
  • Produce crisp, razor-sharp resolution 5kDa - 250kDa
  • Tris-Glycine buffer - Traditional Laemmli separation
  • Marked sample lanes for easy loading and a simple twist-open design
  • 8+1- and 17-well configurations are compatible with multichannel pipetters
  • 2D-well configuration is designed for antibody testing and two-dimensional electrophoresis using a 7cm IPG strip

  • Fit most vertical minigel boxes
  • Packaged in a unique easy-to-open gold plastic cassette
  • Available with a variety of polyacrylamide concentrations and well configurations

Certifications

ISO 9001:2000

Specifications

Specifications

Clear
3.9 in.
10 cm
2° to 8°C
3.9 in.
10 cm
Polyacrylamide
5 to 200 kd
Tris-Glycine
0.55 cm
Protein
10/Pk.
Plastic
Gradient Page
2D Electrophoresis, Denaturing and Native Protein Electrophoresis, Western Blotting
3.9 in.
10 cm
20 μL
3.9 in.
10 cm
Mini
4 to 20%
Denaturing, Native
CBS Scientific MGV System, Daiichi/ISS 2 and 6 System, FisherBiotech Fisher EC 120-2, FisherBiotech Fisher FB-VE 10-1, FisherBiotech Fisher FB-VE 12-1, Hoefer Mighty Small SE 250, Hoefer Mighty Small SE 260, Hoefer Mighty Small SE280, Invitrogen/Novex XCell I and II, Invitrogen/Novex XCell SureLock MiniCell, Lonza PAGEr Minigel Chamber, Owl Separation Systems Penguin P8DS-1, Sigma Aldrich Mini Techware, Zaxis System 200
12-well
2D Well
ISO 9001:2000
3.5 Months
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit