Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ PAGEr™ Gold 4 to 12%, Tris-Glycine, Mini, Protein Gel, 16-well

Separation Range: 25 to 250 kd; Separation Type: Denaturing, Native; For Use With: Biometra Mini V

Manufacturer:  Lonza™ 58524

Catalog No. BMA58524

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PAGEr Gold Precast Gels are easy-to-use protein minigels that offer sharp resolution and consistent protein transfer. Choose from multiple options for denatured or native protein separation over a wide molecular weight range, in an array of well configurations in both 9 x 10cm and 10 x 10cm sizes (see BMA59517 through BMA59524) to fit most popular chambers.

  • Polyacrylamide minigels cast with 4% stacking gel for vertical protein electrophoresis
  • Provide easy, accurate separation of biomolecules over wide molecular-weight range
  • Produce crisp, razor-sharp resolution 5kDa - 250kDa
  • Tris-Glycine buffer - Traditional Laemmli separation
  • Marked sample lanes for easy loading and a simple twist-open design
  • 8+1- and 17-well configurations are compatible with multichannel pipetters

  • Fit most vertical minigel boxes
  • Packaged in a unique easy-to-open gold plastic cassette
  • Available with a variety of polyacrylamide concentrations and well configurations

Certifications

ISO 9001:2000

Specifications

Specifications

2D Electrophoresis, Denaturing and Native Protein Electrophoresis, Western Blotting
3.5 in.
9 cm
14 μL
3.9 in.
10 cm
Mini
4 to 12%
Denaturing, Native
Biometra Mini V, Bio-Rad Min-PROTEAN I, II, and 3, Bio-Rad Ready Gel Cell, CBS Scientific MGV System, FisherBiotech Fisher EC 120-2, Hoefer Mighty Small SE 250, Hoefer Mighty Small SE 260, Hoefer Mighty Small SE280, Invitrogen/Novex XCell I and II, Invitrogen/Novex XCell SureLock MiniCell, Lonza PAGEr Minigel Chamber, Sigma Aldrich Mini Techware
16-well
Plastic
Gradient Page
3.5 in.
9 cm
2° to 8°C
3.9 in.
10 cm
Polyacrylamide
25 to 250 kd
Tris-Glycine
0.49 cm
Protein
10/Pk.
ISO 9001:2000
3.5 Months
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit