Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ PAGEr™ Minigel Chamber and Blot Module System

Designed to provide optimized performance from PAGEr Precast Gels

Manufacturer:  Lonza™ 59905

 View more versions of this product

Catalog No. BMA59905

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Comprises minigel chamber, blot modules and sponge pads
  • Works with most standard precast minigels

Minigel Chamber:

  • Lock-in-place core design with tight, flat fit that eliminates risk of buffer leaks
  • Solid, robust construction
  • Even electrical force ensures straight lanes
  • Runs one or two gels and accommodates a tank blotting module
  • Fits gels of size 9 × 10cm (adapter included) and 10 × 10 cm
  • Buffer volume is approximately 800mL
Blot Module:
  • Works directly in minigel chamber
  • Color-coded cassettes ensure proper orientation of gel during transfer
  • Hinged cassette design for easy assembly
  • Transfer time of 90 min. or less

SDS-PAGE electrophoresis, 2D electrophoresis, Tank blotting

Specifications

Specifications

Protein Electrophoresis
60°C
9 x 10 cm and 10 x 10 cm PAGEr™ Gel
Lower reservoir, Safety cover with attached DC power Leads, Core, 1 blank plate for running single gels, 2 adaptors for running 9cm gels
27.05 oz.
30 w
140°F
150 mA
PAGEr™ Minigel Chamber
250 VDC
800 mL
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit