Learn More
Lonza™ Reliant™ Precast Gels
For rapid and reproducible resolution of DNA sizes from 8bp-10kb - Lonza™ Reliant™ Precast Gels are ideal for analysis of PCR, restriction digest fragments or most cloning needs.
Manufacturer:  Lonza™ 54823
 View more versions of this product
Catalog No. BMA54823
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Ethidium bromide is a powerful mutagen. Gloves should be worn when opening pouches and handling gel solutions containing ethidium bromide.
Description
Description
  • Manufactured with high-quality SeaKem™ Gold and NuSieve™ 3:1 Plus brand agaroses
  • Easy to handle
  • Fit most common mini submerged electrophoresis systems
  • Available with 0.5μg/mL of ethidium bromide for ease of visualizing DNA bands during and after the run
Specifications
Specifications
TBE
4%
12-well
3.7 in.
18° to 26°C
0.5 cm
2.37 in.
Agarose
Yes
12-well
Precast Gel
Clear
DNA separation by gel electrophoresis
4% NuSieve 3:1 Plus Agarose, TBE Buffer, 0.5μg/mL Ethidium Bromide
9.5 cm
0.19 in.
≤15 μL
6 cm
Yes
8 bp to ≥1 kb
20 Gels