Learn More
SeaKem™ HE Agarose, Lonza™
  • For immunoelectrophoresis (IEP), crossed-IEP, counterimmunoelectrophoresis (CEIP), and serum protein electrophoresis
  • High EEO (0.23 –0.26)
  • Standard gelling temperature agarose
    $163.60 - $618.00
Specifications
Chemical Name or Material SeaKem HE Agarose
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BMA50021
View Documents
Lonza Inc
50021
25g
Each for $163.60
Add to cart
BMA50020
View Documents
Lonza Inc
50020
125g
Each for $618.00
Add to cart
Description
Description
Specifications
Specifications
SeaKem HE Agarose