Learn More
SeaKem™ HGT Agarose, Lonza™
  • Used in counterimmunoelectrophoresis and crossed immunoelectrophoresis
  • High gelling temperature (42°C ± 1.5°C)
  • High gel strength (≥1000g/cm2) and clarity
$242.00 - $782.00
Specifications
Chemical Name or Material SeaKem HGT Agarose
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BMA50041
View Documents
Lonza Inc
50041
25g
Each for $242.00
Add to cart
BMA50040
View Documents
Lonza Inc
50040
125g
Each for $782.00
Add to cart
Description
Description
Specifications
Specifications
SeaKem HGT Agarose