Learn More
SeaKem™ ME Agarose, Lonza™
  • For serum protein electrophoresis and nucleic acid electrophoresis
$170.30 - $1642.00
Specifications
Chemical Name or Material SeaKem ME Agarose
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BMA50011
View Documents
Lonza Inc
50011
25g
Each for $170.30
Add to cart
BMA50010
View Documents
Lonza Inc
50010
125g
Each for $563.00
Add to cart
BMA50014
View Documents
Lonza Inc
50014
500g
Each for $1,642.00
Add to cart
Description
Description
Specifications
Specifications
SeaKem ME Agarose