Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lonza™ SimplyLoad™ DNA Ladders
Ready-to-load markers contain loading buffer to save time and pipetting steps
Supplier: Lonza™ 50329
Description
• Consist of RNA transcripts in sizes of 0.5, 1, 1.5, 2, 2.5, 3, 4, 5, 6, and 9kb
• Can be denatured with standard procedures
• Can be visualized on Northern blots with labeled lambda sequence
• Can be detected with ethidium bromide or nucleic acid stains
• Store at -20°C for up to six months or at -80°C for up to 24 months
Specifications
60 ng/μL | |
Liquid | |
Gel electrophoresis | |
16 | |
2° to 8°C |
DNA Molecular Weight Marker/Ladder | |
50% GC content | |
500bp | |
500 μL |
Safety and Handling
ShelfLife : 6 Months
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction