Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Loricrin Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  BiossSupplier Diversity Partner BS18340R

Catalog No. 50-198-4207


Add to cart

Description

Description

This gene encodes loricrin, a major protein component of the cornified cell envelope found in terminally differentiated epidermal cells. Mutations in this gene are associated with Vohwinkel ft.s syndrome and progressive symmetric erythrokeratoderma, both inherited skin diseases.
Specifications

Specifications

Loricrin
Western Blot
Unconjugated
LORICRIN
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
4014
LORICRIN
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
P23490
AI036317; BOS_2784; LOR; Loricrin; LRN; MGC111513; S77319
KLH conjugated synthetic peptide derived from human Loricrin.
Protein A
RUO
Antibody
Polyclonal
Human
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit