Learn More
Globe Scientific Lot Certified Graduated Microcentrifuge Tube
RNase Free, DNase Free, Human DNA Free, Pyrogen Free, ATP Free and PCR Inhibitor Free
$28.80 - $34.00
Specifications
Graduated Yes
Increments 0.5 mL
Max. RCF 20000
No. per Pack 500
Autoclavable Yes
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
22-010-089 View Documents Globe Scientific
111562
1.5 mL Pack of 500 for $28.80
Add to cart
22-010-092 View Documents Globe Scientific
111572
2.0 mL Pack of 500 for $34.00
Add to cart
Description
Description
  • RNase free, DNase free, human DNA free, Pyrogen free, ATP free and PCR inhibitor free
  • Made of polypropylene (PP)
  • With attached leak-resistant snap cap
  • Molded with precise graduation marks for accurate measurements
Specifications
Specifications
Yes
20000
Yes
Natural
0.5 mL
500
Polypropylene
Snap cap