Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOTUS Mouse anti-Human,Mouse,Rat, Unconjugated, Clone: 52-1E, ITM, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen LOTUS
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN25491MI
View Documents
MilliporeSigma
MABN2549100UG
100 μg
Each for $386.00
Add to cart
 
MABN25492MI
View Documents
MilliporeSigma
MABN254925UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

LOTUS Monoclonal antibody specifically detects LOTUS in Human,Mouse,Rat samples. It is validated for Western Blotting.
Specifications

Specifications

LOTUS
Unconjugated
Mouse
Lateral olfactory tract usher substance;Cartilage Acidic Protein–1B
Monoclonal
Protein G Purified
Western Blot
Purified
Human, Mouse, Rat
A recombinant fragment of LOTUS protein consisting of several specific amino acids from the N-terminal region of rat lateral olfactory tract usher substance (LOTUS).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit