Learn More
Lovibond™ Aqua Comparitor Color Discs: Chlorine
For testing water with the Aqua Comparator
Manufacturer:  Lovibond™ 146010
 View more versions of this product
Catalog No. 09-053-139
Add to cart
Description
Description
Economical replacement discs make it easy to test for chlorine using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Specifications
0 to 1mg/L
Aqua comparator™
Testing water with the Aqua comparator™
Chlorine Color Disc