Learn More
Lovibond™ Aqua Comparitor Color Discs: Chlorine
For testing water with the Aqua Comparator
Manufacturer:  Lovibond™ 146030
 View more versions of this product
Catalog No. 09-053-141
Add to cart
Description
Description
Economical replacement discs make it easy to test for chlorine using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Specifications
10 to 300mg/L
Aqua comparator™
Testing water with the Aqua comparator™
Chlorine Color Disc