Learn More
Lovibond™ Aqua Comparitor Color Discs: Copper
For testing water with the Aqua Comparator
Manufacturer:  Lovibond™ 146441
 View more versions of this product
Catalog No. 09-053-174
Add to cart
Description
Description
Economical replacement discs make it easy to test for copper using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Specifications
0.01 to 0.2mg/L
Aqua comparator™
Testing water with the Aqua comparator™
Copper LR Powder Color Disc