Learn More
Lovibond™ Aqua Comparitor Color Discs: Manganese
For testing water with the Aqua Comparator
Supplier: Lovibond™ 146410
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for manganese using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ | |
0.1 to 0.7mg/L | |
Manganese LR Color Disc |