Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Lovibond™ Aqua Comparitor Color Discs: Manganese
For testing water with the Aqua Comparator
Manufacturer:  Lovibond™ 146420
 View more versions of this product
Catalog No. 09-053-170
Description
Description
Economical replacement discs make it easy to test for manganese using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Specifications
0.02 to 0.2mg/L
Aqua comparator™
Testing water with the Aqua comparator™
Manganese VLR Color Disc