Learn More
Lovibond™ Aqua Comparitor Color Discs: Molybdate
For testing water with the Aqua Comparator
$38.25
Specifications
For Use With (Application) Testing water with the Aqua comparator™
For Use With (Equipment) Aqua comparator™
View More Specs
Products 2
Catalog Number Mfr. No. Type Range Price Quantity & Availability  
Catalog Number Mfr. No. Type Range Price Quantity & Availability  
09-053-156
View Documents
Lovibond™
146290
Molybdate HR Color Disc 0 to 100mg/L
Each for $38.25
Add to Cart
09-053-157
View Documents
Lovibond™
146291
Molybdate LR Color Disc 0 to 10mg/L
Each for $38.25
Add to Cart
Description
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for molybdate using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Specifications
Testing water with the Aqua comparator™
Aqua comparator™