Learn More
Lovibond™ Aqua Comparitor Color Discs: pH
For testing water with the Aqua Comparator
Manufacturer:  Lovibond™ 146100
 View more versions of this product
Catalog No. 09-053-144
Add to cart
Description
Description
Economical replacement discs make it easy to test for pH using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Specifications
6.5 to 8.4 pH
Aqua comparator™
Testing water with the Aqua comparator™
pH Color Disc