Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Lovibond™ Borosilicate Glass Colorimetry Cells
Precision fused cells
Manufacturer:  Lovibond™ 656130
 View more versions of this product
Catalog No. 09-053-355
Description
Description
Available in a range of dimensions and path lengths, made from optical glass to the highest standards. Note: For instruments equipped with a heater unit and whenever cells are subjected to thermal shock, it is recommended that borosilicate cells be used.
Series: W600 Borosilicate
Specifications
Specifications
Model Fs, PFXi-880, 950 and 995 instruments with heated samples (up to 100°C)
Borosilicate Glass
13.33cm
Colorimetry Cell