Lovibond™ CHECKIT™ Comparator Test Kits

Accurate test kits for routine water analysis

Manufacturer: Lovibond™ 147430

 View all options for this product

Catalog No. 09-053-113

  Check Availability
Add to Cart


    Range 0-5.0mg/L
    For Use With (Application) For routine water analysis
    Product Type Copper
    Sufficient For 100 tests

    • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
    • Color discs and reagents are available separately

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit