Learn More
Lovibond™ COD Tube Tests; MR; Mercury Free
Premeasured unit dose reagent for water quality testing
Manufacturer:  Lovibond™ 2420711
 View more versions of this product
Catalog No. 09-053-311
Add to cart
Description
Description
  • For use in SC400™ Colorimeters and Aqua Comparator™ Test Kits
  • Also suitable for use in Hach™ instruments and test kits
  • Easy-to-open, sealed vials eliminate risk of cross-contamination
  • Extended shelf life of up to five years
For Municipal Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted test method
Specifications
Specifications
US EPA
Municipal wastewater testing, industrial wastewater and process control testing, environmental water testing
25.0
0 to 1500mg/L
SC400 colorimeters and Aqua comparator™ test kits, Hach™ instruments
Dichromate/H2SO4
Safety and Handling
Safety and Handling
ShelfLife : 5 Years