Learn More
Lovibond™ Color of Varnish Discs for Lovibond™ 605 and 705 Comparator Bodies
For measuring Gardner (Varnish) color scale
Supplier: Lovibond™ 296120
This item is not returnable.
View return policy
Description
For Lovibond 605 and 705 Comparator Bodies (using 10 mm pathlength) for measuring Gardner (Varnish) color scale.
- Ranges 1 to 9 or 9 to 18, in steps of one unit
- Utilizes 1933 Gardner Method
Specifications
Color of Varnish Disc (9.0 to 18.0) | |
For measuring Gardner (Varnish) color scale | |
Orbeco 605 and 705 comparator bodies |